missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ricca Chemical Company Sodium Thiosulfate, 0.0100 N (N/100), Ricca Chemical
Supplier: Ricca Chemical Company 78901
Specifications
SODIUM THIOSULFATE | |
0.2,98.0,0.0016 | |
H10Na2O8S2 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
disodium pentahydrate sulfanidesulfonate | |
61475 | |
Natural Poly Bottle |
10102-17-7,7732-18-5,497-19-8 | |
0.21,100.0,0.0016 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O | |
248.17 | |
CHEBI:32150 | |
4 L |
Chemical Identifiers
10102-17-7 | |
248.17 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
CHEBI:32150 | |
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O |
H10Na2O8S2 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
61475 | |
disodium pentahydrate sulfanidesulfonate |