missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium thiophosphate hydrate

Manufacturer:  Alfa Aesar™ 5614614

 View more versions of this product

Catalog No. AA5614614



Specifications

Specifications

Sodium thiophosphate hydrate
H2Na3O4PS
14,8693
VYYYFZBRAKXIPU-UHFFFAOYSA-K
trisodium;thiophosphate;hydrate
53442939
Crystalline Aggregates
53°C to 58°C
10489-48-2
MFCD00041513
sodium thiophosphate hydrate, sodium phosphorothioate hydrate, sodium thiophosphate tribasic hydrate, trisodium hydrate thiophosphate, trisodium hydrate phosphorothioate, sodium phosphorothioate-water 3/1/1, sodium thiophosphate tribasic: hydrate
O.[O-]P(=S)([O-])[O-].[Na+].[Na+].[Na+]
198.015
179.93 (Anhydrous)
Green to White
25g

Chemical Identifiers

10489-48-2
198.015
VYYYFZBRAKXIPU-UHFFFAOYSA-K
53442939
O.[O-]P(=S)([O-])[O-].[Na+].[Na+].[Na+]
H2Na3O4PS
MFCD00041513
sodium thiophosphate hydrate, sodium phosphorothioate hydrate, sodium thiophosphate tribasic hydrate, trisodium hydrate thiophosphate, trisodium hydrate phosphorothioate, sodium phosphorothioate-water 3/1/1, sodium thiophosphate tribasic: hydrate
trisodium;thiophosphate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents