missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium thiophenolate, 90%, Thermo Scientific™

Chemical Identifiers

CAS 930-69-8
Molecular Formula C6H5NaS
Molecular Weight (g/mol) 132.16
MDL Number MFCD00066460
InChI Key RZWQDAUIUBVCDD-UHFFFAOYSA-M
Synonym sodium thiophenolate, sodium thiophenoxide, sodium benzenethiolate, benzenethiol, sodium salt, sodium thiophenylate, sodium phenylthiolate, thiophenol sodium salt, unii-75gf5o2o8y, benzenethiol sodium salt, benzenethiol, sodium salt 1:1
PubChem CID 4110039
IUPAC Name sodium;benzenethiolate
SMILES [Na+].[S-]C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC436260100
SDS View Documents Promotion Details
Thermo Scientific Chemicals
436260100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC436260500
SDS View Documents Promotion Details
Thermo Scientific Chemicals
436260500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

930-69-8
132.16
RZWQDAUIUBVCDD-UHFFFAOYSA-M
4110039
[Na+].[S-]C1=CC=CC=C1
C6H5NaS
MFCD00066460
sodium thiophenolate, sodium thiophenoxide, sodium benzenethiolate, benzenethiol, sodium salt, sodium thiophenylate, sodium phenylthiolate, thiophenol sodium salt, unii-75gf5o2o8y, benzenethiol sodium salt, benzenethiol, sodium salt 1:1
sodium;benzenethiolate

Specifications

930-69-8
88% min. (Iodimetry)
C6H5SNa
sodium thiophenolate, sodium thiophenoxide, sodium benzenethiolate, benzenethiol, sodium salt, sodium thiophenylate, sodium phenylthiolate, thiophenol sodium salt, unii-75gf5o2o8y, benzenethiol sodium salt, benzenethiol, sodium salt 1:1
RZWQDAUIUBVCDD-UHFFFAOYSA-M
sodium;benzenethiolate
4110039
90%
Glass bottle
10g
C6H5NaS
MFCD00066460
Solubility in water: soluble
[Na+].[S-]C1=CC=CC=C1
132.16
132.16
Authentic
Sodium thiophenolate
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently

GHS Signal Word: Danger

EINECSNumber : 213-224-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only