missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium thiophenolate, 90%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC436260100
Specifications
Sodium thiophenolate | |
88% min. (Iodimetry) | |
C6H5SNa | |
Solubility in water: soluble | |
C1=CC=C(C=C1)[S-].[Na+] | |
132.16 | |
132.16 | |
90% | |
Glass bottle | |
Beige to White | |
10g |
930-69-8 | |
C6H5NaS | |
benzenethiol sodium salt, benzenethiol, sodium salt, benzenethiol, sodium salt 1:1, sodium benzenethiolate, sodium phenylthiolate, sodium thiophenolate, sodium thiophenoxide, sodium thiophenylate, thiophenol sodium salt, unii-75gf5o2o8y | |
RZWQDAUIUBVCDD-UHFFFAOYSA-M | |
sodium;benzenethiolate | |
4110039 | |
Powder | |
Authentic | |
Solubility in water: soluble. | |
>300.0°C |
Chemical Identifiers
930-69-8 | |
132.16 | |
benzenethiol sodium salt, benzenethiol, sodium salt, benzenethiol, sodium salt 1:1, sodium benzenethiolate, sodium phenylthiolate, sodium thiophenolate, sodium thiophenoxide, sodium thiophenylate, thiophenol sodium salt, unii-75gf5o2o8y | |
sodium;benzenethiolate |
C6H5NaS | |
RZWQDAUIUBVCDD-UHFFFAOYSA-M | |
4110039 | |
C1=CC=C(C=C1)[S-].[Na+] |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently
GHS Signal Word: Danger
EINECSNumber : 213-224-