Learn More
Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 305710010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium thiomethoxide | |
White | |
94 to 106% (Iodimetry) | |
CH3NaS | |
MFCD00174316 | |
sodium thiomethoxide, sodium methanethiolate, sodium thiomethylate, methyl mercaptan sodium salt, methanethiol sodium salt, sodiumthiomethoxide, methanethiol, sodium salt, sodiummethanethiolate, sodium methyl mercaptide, methylsulfanyl sodium | |
RMBAVIFYHOYIFM-UHFFFAOYSA-M | |
sodium;methanethiolate | |
4378561 | |
95% | |
Powder |
5188-07-8 | |
Authentic | |
Glass bottle | |
CH3SNa | |
1 g | |
Solubility in water: freely soluble. | |
C[S-].[Na+] | |
70.09 | |
70.09 | |
Pure |
Chemical Identifiers
5188-07-8 | |
70.09 | |
RMBAVIFYHOYIFM-UHFFFAOYSA-M | |
C[S-].[Na+] |
CH3NaS | |
MFCD00174316 | |
4378561 |
Safety and Handling
GHS H Statement
Flammable solid.
Toxic if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
EINECSNumber : 225-969-9
RUO – Research Use Only