Please login to your online account to display your discounted pricing

Sodium Thiocyanate, Certified, 0.1000N ±0.0005N (0.1M), LabChem™

$54.80 - $215.60

Chemical Identifiers

Chemical Name or Material Sodium Thiocyanate
CAS 7732-18-5
CAS Max % 99.2
CAS 540-72-7
CAS Max % 0.8
Purity Grade Certified Grade
Linear Formula NaSCN
Formula Weight 81.07
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC249701

 
lab chem
LC249701
500mL Each for $54.80

LC249702

 
lab chem
LC249702
1L Each for $82.17

LC249704

 
lab chem
LC249704
4L Each for $215.60
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 81.07
Chemical Name or Material Sodium Thiocyanate
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Linear Formula NaSCN
CAS 7732-18-5
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Sulfur compounds
Color Clear, colorless
Quantity 500mL
CAS Max % 99.2
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 540-72-7
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.8
Concentration 0.1000N ±0.0005N (0.1M)
Density 1g/mL
Traceability to NIST Traceable to NIST
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.