Please login to your online account to display your discounted pricing

Sodium Thiocyanate, ACS Grade, ≥ 98.0%, LabChem™

$174.94 - $174.94

Chemical Identifiers

Chemical Name or Material Sodium Thiocyanate
CAS 540-72-7
CAS Max % 100
Purity Grade ACS Grade
Linear Formula NaSCN
Formula Weight 81.07
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC249601

 
lab chem
LC249601
500g Each for $174.94
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 81.07
Specific Gravity 1.74g/cm3
Chemical Name or Material Sodium Thiocyanate
EINECS Number 208-754-4
Identification Passes Test
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystalline
Solubility Soluble in water
Storage Information Room Temperature
Linear Formula NaSCN
CAS 540-72-7
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Sulfur compounds
Color White
Melting Point 287°C
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Harmful if swallowed, in contact with skin or if inhaled.
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
Density 1.74g/cm3
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed, in contact with skin or if inhaled.