Learn More
Sodium thiocyanate, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A118670I
Description
A compound used to convert alkyl halides into the corresponding alkylthiocyanates. Undergoes nucleophilic substitution reactions with phase-transfer catalysts. Silver thiocyanate may be used as well, the precipitation of insoluble silver halides help simplify workup. Treatment of isopropyl bromide with sodium thiocyanate in a hot ethanolic solution affords isopropyl thiocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA compound used to convert alkyl halides into the corresponding alkylthiocyanates. Undergoes nucleophilic substitution reactions with phase-transfer catalysts. Silver thiocyanate may be used as well, the precipitation of insoluble silver halides help simplify workup. Treatment of isopropyl bromide with sodium thiocyanate in a hot ethanolic solution affords isopropyl thiocyanate.
Solubility
Soluble in water (1250g/L), alcohol, and acetone.
Notes
Hygroscopic, protect from humidity and water. Store in cool, dry conditions in well sealed container.
Specifications
Sodium thiocyanate | |
287°C | |
6 to 8 | |
CNNaS | |
5000 g | |
Hygroscopic; Light sensitive | |
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased | |
VGTPCRGMBIAPIM-UHFFFAOYSA-M | |
sodium;thiocyanate | |
516871 | |
81.07 |
540-72-7 | |
1.74 | |
Odorless | |
MFCD00011123 | |
3594965 | |
14,9327 | |
Soluble in water (1250g/L),alcohol,and acetone. | |
[Na+].[S-]C#N | |
81.07 | |
CHEBI:30952 | |
96% |
Chemical Identifiers
540-72-7 | |
81.07 | |
VGTPCRGMBIAPIM-UHFFFAOYSA-M | |
516871 | |
sodium;thiocyanate |
CNNaS | |
MFCD00011123 | |
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased | |
CHEBI:30952 | |
[Na+].[S-]C#N |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P302+P352-P304+P340-P314-P330-P362-P501c
H302+H312+H332-H373
EINECSNumber : 208-754-4
RTECSNumber : XL2275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only