missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium thiocyanate, 96%

Manufacturer:  Alfa Aesar™ A1186730

 View more versions of this product

Catalog No. AAA1186730



Description

Description

Applications
A compound used to convert alkyl halides into the corresponding alkylthiocyanates. Undergoes nucleophilic substitution reactions with phase-transfer catalysts. Silver thiocyanate may be used as well, the precipitation of insoluble silver halides help simplify workup. Treatment of isopropyl bromide with sodium thiocyanate in a hot ethanolic solution affords isopropyl thiocyanate.

Solubility
Soluble in water (1250g/L), alcohol, and acetone.

Notes
Hygroscopic, protect from humidity and water. Store in cool, dry conditions in well sealed container.
Specifications

Specifications

Sodium thiocyanate
287°C
6 to 8
CNNaS
3594965
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
C(#N)[S-].[Na+]
81.068
CHEBI:30952
96%
Odorless
250g
540-72-7
MFCD00011123
14,9327
VGTPCRGMBIAPIM-UHFFFAOYSA-M
sodium;thiocyanate
516871
81.07

Chemical Identifiers

540-72-7
81.068
VGTPCRGMBIAPIM-UHFFFAOYSA-M
516871
sodium;thiocyanate
CNNaS
MFCD00011123
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
CHEBI:30952
C(#N)[S-].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XL2275000

TSCA : TSCA

SDS Documents