missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tetrakis(4-fluorophenyl)borate dihydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5595206

 View more versions of this product

Catalog No. AAH5595206



Description

Description

Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents.
Specifications

Specifications

Sodium tetrakis(4-fluorophenyl)borate dihydrate
C24H20BF4NaO2
5 g
sodium tetrakis 4-fluorophenyl borate dihydrate, cesibor, tetrakis 4-fluorophenyl boron sodium, sodium tetrakis 4-fluorophenyl boranuide dihydrate, sodium tetrakis 4-fluorophenyl borate dihydrate nt, sodium tetrakis 4-fluorophenyl borate dihydrate, selectophore tm
[B-](C1=CC=C(C=C1)F)(C2=CC=C(C=C2)F)(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.O.O.[Na+]
450.215
450.21 (414.18 Anhydrous)
207683-22-5
MFCD00149598
4285808
MSDGDEJOIBMWJD-UHFFFAOYSA-N
sodium;tetrakis(4-fluorophenyl)boranuide;dihydrate
45073643
98%

Chemical Identifiers

207683-22-5
450.215
MSDGDEJOIBMWJD-UHFFFAOYSA-N
45073643
[B-](C1=CC=C(C=C1)F)(C2=CC=C(C=C2)F)(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.O.O.[Na+]
C24H20BF4NaO2
MFCD00149598
sodium tetrakis 4-fluorophenyl borate dihydrate, cesibor, tetrakis 4-fluorophenyl boron sodium, sodium tetrakis 4-fluorophenyl boranuide dihydrate, sodium tetrakis 4-fluorophenyl borate dihydrate nt, sodium tetrakis 4-fluorophenyl borate dihydrate, selectophore tm
sodium;tetrakis(4-fluorophenyl)boranuide;dihydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only