missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Tetrakis[3,5-bis(trifluoromethyl)phenyl]borate Hydrate 98.0+%, TCI America™

Manufacturer:  TCI America S04471G

 View more versions of this product

Catalog No. S04471G



Specifications

Specifications

Sodium Tetrakis[3,5-bis(trifluoromethyl)phenyl]borate Hydrate
1g
C32H12BF24Na
nabarf
[B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+]
886.209
886.21
≥98.0% (HPLC,W)
Undesignated
79060-88-1
MFCD00043323
LTGMONZOZHXAHO-UHFFFAOYSA-N
sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide
23681909
Crystal-Powder at 20°C

Chemical Identifiers

79060-88-1
886.209
LTGMONZOZHXAHO-UHFFFAOYSA-N
23681909
[B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+]
C32H12BF24Na
MFCD00043323
nabarf
sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide
Safety and Handling

Safety and Handling

TSCA : No

Documents