Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium tetrakis(1-imidazolyl)borate, 97%

Chemical Identifiers

Chemical Name or Material Sodium tetrakis(1-imidazolyl)borate
CAS 68146-65-6
Formula Weight 302.08
MDL Number MFCD00012083
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5508103

  SDS
alfa aesar™
H5508103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5508106

  SDS
alfa aesar™
H5508106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5508114

  SDS
alfa aesar™
H5508114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 302.08
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Chemical Name or Material Sodium tetrakis(1-imidazolyl)borate
CAS 68146-65-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Physical Form Solid
Quantity 1g
Health Hazard 1 Warning
MDL Number MFCD00012083
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.