Please login to your online account to display your discounted pricing

Sodium tetrafluoroborate, 98%, extra pure, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC192120025

 View more versions of this product

Catalog No. AC192120025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.0% (Titrimetry other)
Formula Weight 109.79
Physical Form Powder, Crystals or Needles
Purity Grade Extra Pure
Quantity 2.5kg
Color White to yellow to beige to brown
TSCA TSCA
EINECS Number 237-340-6
RTECS Number ED2975000
Density 2.47
Merck Index 15, 8753
Packaging Plastic bottle
Sulfate 1000ppm max.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
CAS 13755-29-8
Molecular Formula BF4Na
Name Note 98%
MDL Number MFCD00003515
Chemical Name or Material Sodium tetrafluoroborate
Assay Percent Range 97+%
Linear Formula NaBF4
Iron (Fe) 30ppm max.
Chlorine (Cl) 1000ppm max.
Chloride 200ppm max.
Heavy Metals (as Pb) 30ppm max.
Synonym Sodium fluoborate
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger

Specifications

Chemical Name or Material Sodium tetrafluoroborate
Name Note 98%
CAS 13755-29-8
Purity Grade Extra Pure
Assay ≥97.0% (Titrimetry other)
Assay Percent Range 97+%
Molecular Formula BF4Na
Linear Formula NaBF4
Formula Weight 109.79
MDL Number MFCD00003515
Synonym Sodium fluoborate
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents