missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A12393A3

 View more versions of this product

Catalog No. AAA12393A3



Description

Description

Sodium tetrafluoroborate is used in fluxes, metal finishing and in plating circuits. It is used as a starting material in the preparation of ionic liquids as well as boron trifluoride. It acts as an efficient catalyst for the synthesis of 3,4-dihydropyrimidin-2(1H)-ones and thiones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tetrafluoroborate is used in fluxes, metal finishing and in plating circuits. It is used as a starting material in the preparation of ionic liquids as well as boron trifluoride. It acts as an efficient catalyst for the synthesis of 3,4-dihydropyrimidin-2(1H)-ones and thiones.

Solubility
Soluble in water.

Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

Sodium tetrafluoroborate
13755-29-8
BF4Na
MFCD00003515
sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
KGJZTOFHXCFQIV-UHFFFAOYSA-N
sodium;tetrafluoroborate
4343483
97%
2.47
384°C (dec.)
2 kg
NaBF4
14,8617
Soluble in water.
[Na+].F[B-](F)(F)F
109.79
109.79
Hygroscopic

Chemical Identifiers

13755-29-8
109.79
KGJZTOFHXCFQIV-UHFFFAOYSA-N
4343483
[Na+].F[B-](F)(F)F
BF4Na
MFCD00003515
sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
sodium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 237-340-6

RTECSNumber : ED2975000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only