Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium tetraethylborate, 98%

Chemical Identifiers

Chemical Name or Material Sodium tetraethylborate
CAS 15523-24-7
Molecular Formula C8H20BNa
Linear Formula NaB(C2H5)4
Formula Weight 150.05
MDL Number MFCD00061547
UN Number UN3393
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8913703

  SDS
alfa aesar
8913703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA8913706

  SDS
alfa aesar™
8913706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 150.05
Quantity 1g
Assay 98%
Linear Formula NaB(C2H5)4
UN Number UN3393
Solubility Information Dissolves in water with decomposition
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H250-H260-H252
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
Health Hazard 3 GHS P Statement
P210-P222-P231+P232-P370+P378c-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not allow contact with air.
Handle under inert gas.
Protect from moisture.
In case of fire:
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC, WATER-REACTIVE
Chemical Name or Material Sodium tetraethylborate
Physical Form Crystalline
Assay Percent Range 98%
Molecular Formula NaB(C2H5)4
MDL Number MFCD00061547
CAS 15523-24-7
Safety and Handling

Danger

GHS P Statement
P210-P222-P231+P232-P370+P378c-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not allow contact with air.
Handle under inert gas.
Protect from moisture.
In case of fire:

GHS H Statement
H250-H260-H252
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.