missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tetrachloroplatinate(II) hydrate, Premion™, 99.95% (metals basis), Pt 42.4% min, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01216803

 View more versions of this product

Catalog No. AA1216803



Description

Description

Sodium tetrachloroplatinate(II) hydrate is used as chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium tetrachloroplatinate(II) hydrate
207683-21-4
1 g
Cl4Na2Pt
sodium tetrachloroplatinate ii hydrate, disodium tetrachloroplatinate 2-hydrate, sodium tetrachloroplatinate 2--water 2/1/1
DOAIHRYZKUCMDX-UHFFFAOYSA-J
disodium;tetrachloroplatinum(2-);hydrate
56846332
99.95%
Pt 42.4% min.
100°C
Crystalline
(metals basis)
MFCD00150188
Soluble in water.
[Na+].[Na+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt--]
382.87
382.88 (Anhydrous)
Premion™

Chemical Identifiers

207683-21-4
382.87
DOAIHRYZKUCMDX-UHFFFAOYSA-J
56846332
[Na+].[Na+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt--]
Cl4Na2Pt
MFCD00150188
sodium tetrachloroplatinate ii hydrate, disodium tetrachloroplatinate 2-hydrate, sodium tetrachloroplatinate 2--water 2/1/1
disodium;tetrachloroplatinum(2-);hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H300-H334-H318-H315-H317
Fatal if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c

H302+H312+H332-H315-H317-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 233-051-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only