missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium tetrachloroplatinate(II) hydrate, Premion™, 99.95% (metals basis), Pt 42.4% min

Manufacturer:  Alfa Aesar™ 1216806

 View more versions of this product

Catalog No. AA1216806



Specifications

Specifications

Sodium tetrachloroplatinate(II) hydrate
(metals basis)
MFCD00150188
GIEPPPFPHULPBK-UHFFFAOYSA-J
disodium;tetrachloroplatinum(2-);hydrate
56846332
Crystalline
Premion™
100°C
207683-21-4
Cl4H2Na2OPt
sodium tetrachloroplatinate ii hydrate, disodium tetrachloroplatinate 2-hydrate, sodium tetrachloroplatinate 2--water 2/1/1
O.[Na+].[Na+].Cl[Pt-2](Cl)(Cl)Cl
400.879
382.88 (Anhydrous)
99.95%
Pt 42.4% min.
5g

Chemical Identifiers

207683-21-4
400.879
GIEPPPFPHULPBK-UHFFFAOYSA-J
56846332
O.[Na+].[Na+].Cl[Pt-2](Cl)(Cl)Cl
Cl4H2Na2OPt
MFCD00150188
sodium tetrachloroplatinate ii hydrate, disodium tetrachloroplatinate 2-hydrate, sodium tetrachloroplatinate 2--water 2/1/1
disodium;tetrachloroplatinum(2-);hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H300-H334-H318-H315-H317
Fatal if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents