missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium tetrachloropalladate(II) hydrate, Premion™, 99.999% (metals basis), Pd 30% min

Manufacturer:  Alfa Aesar™ 4394506

 View more versions of this product

Catalog No. AA4394506



Specifications

Specifications

Sodium tetrachloropalladate (II) hydrate
(metals basis)
Na2PdCl4·xH2O (x≈3)
sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
[Na+].[Na+].Cl[Pd-2](Cl)(Cl)Cl
294.2
294.20 (Anhydrous)
99.999%
Pd 30% min
Red to Brown
13820-53-6
Cl4Na2Pd
MFCD00003487
ZIXLZKBNIAXVBE-UHFFFAOYSA-J
disodium;tetrachloropalladium(2-)
11000870
Crystalline
Premion™
Hygroscopic
5g

Chemical Identifiers

13820-53-6
294.2
ZIXLZKBNIAXVBE-UHFFFAOYSA-J
11000870
[Na+].[Na+].Cl[Pd-2](Cl)(Cl)Cl
Cl4Na2Pd
MFCD00003487
sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
disodium;tetrachloropalladium(2-)
Safety and Handling

Safety and Handling

GHS H Statement
H318-H290-H302
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.

GHS P Statement
P280f-P305+P351+P338
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Danger

RTECSNumber : RT3480000

TSCA : TSCA

Documents