missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tetrachloroaurate(III) dihydrate, 99.99% (metals basis), Alfa Aesar™, Premion™

Manufacturer:  Alfa Aesar 1214806

 View more versions of this product

Catalog No. AA1214806



Specifications

Specifications

Sodium tetrachloroaurate(III) dihydrate
99.99% (metals basis)
MFCD00149162
OEERDTKOVZFOHF-UHFFFAOYSA-J
sodium;tetrachlorogold(1-);dihydrate
18949434
Crystalline
5g
13874-02-7
AuCl4H4NaO2
sodium tetrachloroaurate iii dihydrate, sodium tetrachloroaurate iii dihydrate, sodium;tetrachlorogold 1-;dihydrate, sodiumtetrachloroaurate iii dihydrate, sodium dihydrate tetrachloroaurate
O.O.[Na+].Cl[Au-](Cl)(Cl)Cl
397.786
397.80 (361.77 Anhydrous)
0.8g/mL

Chemical Identifiers

13874-02-7
397.786
OEERDTKOVZFOHF-UHFFFAOYSA-J
18949434
O.O.[Na+].Cl[Au-](Cl)(Cl)Cl
AuCl4H4NaO2
MFCD00149162
sodium tetrachloroaurate iii dihydrate, sodium tetrachloroaurate iii dihydrate, sodium;tetrachlorogold 1-;dihydrate, sodiumtetrachloroaurate iii dihydrate, sodium dihydrate tetrachloroaurate
sodium;tetrachlorogold(1-);dihydrate
Safety and Handling

Safety and Handling

EINECSNumber : 239-241-3

Documents