Please login to your online account to display your discounted pricing

Sodium tetrachloroaurate hydrate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Sodium tetrachloroaurate hydrate
Assay 48.3 to 50.3% (Au)
Assay Percent Range 98%
Molecular Formula AuCl4Na·xH2O
Linear Formula NaAuCl4·xH2O
Formula Weight 361.77
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC396270050

  SDS
thermo fisher scientific
AC396270050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC396270010

  SDS
thermo fisher scientific
AC396270010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder and/or Lumps
Assay 48.3 to 50.3% (Au)
Formula Weight 361.77
Quantity 5g
Color Yellow to orange to dark brown
Chemical Name or Material Sodium tetrachloroaurate hydrate
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula AuCl4Na·xH2O
Linear Formula NaAuCl4·xH2O
CAS 29156-65-8
Health Hazard 1 Warning
Safety and Handling

Warning