Learn More
Sodium tetrachloroaluminate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02237514
Description
Sodium tetrachloroaluminate is used as Friedel-Crafts acylating agent , dehydrating and dehydrogenating agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium tetrachloroaluminate | |
7784-16-9 | |
Powder | |
AlCl4Na | |
sodium tetrachloroaluminate, sodium tetrachloroaluminate 1-, alcl4.na, aluminum sodium chloride, naalcl4, sodium tetrachloroaluminate, anhydrous, powder trace metals basis | |
CMLRNXGMTZKKSR-UHFFFAOYSA-J | |
sodium;tetrachloroalumanuide | |
16699350 | |
Moisture sensitive | |
2.01 g/cm3 |
185°C | |
Yellow | |
25 g | |
MFCD00050361 | |
Soluble in water | |
[Na+].[Al-](Cl)(Cl)(Cl)Cl | |
191.771 | |
191.78 | |
Pale yellow powder |
Chemical Identifiers
7784-16-9 | |
191.771 | |
CMLRNXGMTZKKSR-UHFFFAOYSA-J | |
16699350 | |
[Na+].[Al-](Cl)(Cl)(Cl)Cl |
AlCl4Na | |
MFCD00050361 | |
sodium tetrachloroaluminate, sodium tetrachloroaluminate 1-, alcl4.na, aluminum sodium chloride, naalcl4, sodium tetrachloroaluminate, anhydrous, powder trace metals basis | |
sodium;tetrachloroalumanuide |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8, Packing Group: III
EINECSNumber : 232-050-6
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only