missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Tetraborate Decahydrate (Powder or Granular/NF), Fisher Chemical

Manufacturer:  Fisher Scientific Company FLS24612

 View more versions of this product

Catalog No. S246-12


 

Specifications Description & Specifications

Specifications

Sodium Tetraborate Decahydrate
NF
99 to 105%
B4Na2O7 · 10 H2O
MFCD00149193
Pass Test
0.002% max.
Fiber Drum
White
9
1303-96-4
Powder/Solid
99 to 105 %
381.37
Borax
1.7300g/cm³
Pass Test
Meets Requirements
75°C
12kg

Chemical Identifiers

1303-96-4
99 to 105 %
381.37
Borax
99 to 105%
Powder/Solid
B4Na2O7 · 10 H2O
MFCD00149193
Sodium Tetraborate Decahydrate
NF

Safety and Handling

SDS & Safety Documents