missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium tetraborate decahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 205950050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysisSpecifications
Sodium tetraborate decahydrate | |
White | |
9 to 9.5 (5% soln. at 20°C) | |
5 kg | |
B4Na2O7·10H2O | |
MFCD00149193 | |
15,8726 | |
Solubility in water: 60g/L (20°C). Other solubilities: soluble in glycerol,insoluble in alcohol | |
381.36 | |
Analytical |
75.0°C | |
1303-96-4 | |
Fine Crystalline Powder | |
99.5% | |
Na2B4O7·10H2O | |
04,461 | |
Borax, Sodium borate decahydrate | |
381.36 | |
99.5% | |
Plastic drum |
Chemical Identifiers
1303-96-4 | |
381.36 | |
Borax, Sodium borate decahydrate |
B4Na2O7·10H2O | |
MFCD00149193 |
Safety and Handling
GHS H Statement
May damage fertility.
May damage the unborn child.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
RUO – Research Use Only