missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tetraborate decahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 205950010

 View more versions of this product

Catalog No. AC205950010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for analysis
Specifications

Specifications

Sodium tetraborate decahydrate
White
9 to 9.5 (5% soln. at 20°C)
1 kg
B4Na2O7·10H2O
MFCD00149193
15,8726
Solubility in water: 60g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
381.36
Analytical
75.0°C
1303-96-4
Fine Crystalline Powder
99.5%
Na2B4O7·10H2O
04,461
Borax, Sodium borate decahydrate
381.36
99.5%
Plastic Bottle

Chemical Identifiers

1303-96-4
381.36
Borax, Sodium borate decahydrate
B4Na2O7·10H2O
MFCD00149193
Safety and Handling

Safety and Handling

GHS H Statement
May damage fertility.
May damage the unborn child.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only