Learn More
Sodium tetraborate, anhydrous, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01087514
Description
Sodium tetraborate is used as a co-catalyst for (2,2,6,6-tetramethylpiperidin-1-yl)oxyl and sodium hypochlorite oxidation of alcohols in greener and non-chlorinated solvents. It acts as a reagent for the decomposition of oxides and silicates by fusion. It is used to prepare buffers, which is used in gel electrophoresis of DNA. It acts as a source of borate, micronutrient fertilizer, preservative in taxidermy, additive in ceramic slips and glazes. It is also used as a precursor for sodium perborate monohydrate and boric acid. It finds application as fluoride detoxification, neutron absorber in nuclear reactors and to make indelible ink for dip pens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1575°C (decomposition) | |
741°C | |
1330-43-4 | |
Powder | |
(metals basis) | |
Na2B4O7 | |
14,8590 | |
Soluble in water and glycerol. Slightly soluble in ethanol. Insoluble in acids. | |
[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 | |
201.21 | |
99.998% | |
Odorless | |
Hygroscopic |
Sodium tetraborate | |
Anhydrous | |
White | |
25 g | |
B4Na2O7 | |
MFCD00081185 | |
Borax, anhydrous; Sodium borate (tetra) | |
NXSVMNYRVBQCMO-UHFFFAOYSA-N | |
disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) | |
201.22 | |
Puratronic™ | |
1.501 | |
2.367 g/mL |
Chemical Identifiers
1330-43-4 | |
201.21 | |
NXSVMNYRVBQCMO-UHFFFAOYSA-N | |
disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) |
B4Na2O7 | |
MFCD00081185 | |
Borax, anhydrous; Sodium borate (tetra) | |
[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
H319-H360FD
EINECSNumber : 215-540-4
RTECSNumber : ED4588000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only