missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tert-Pentoxide 97.0+%, TCI America™

Manufacturer:  TCI America S0950100G

 View more versions of this product

Catalog No. S0950100G



Specifications

Specifications

Sodium tert-Pentoxide
100g
C5H11NaO
3206
CGRKYEALWSRNJS-UHFFFAOYSA-N
sodium;2-methylbutan-2-olate
2733478
Crystal-Powder at 20°C
White-Yellow
14593-46-5
MFCD00064245
sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt
CCC(C)(C)[O-].[Na+]
110.132
110.13
≥97.0% (T)

Chemical Identifiers

14593-46-5
110.132
CGRKYEALWSRNJS-UHFFFAOYSA-N
2733478
CCC(C)(C)[O-].[Na+]
C5H11NaO
MFCD00064245
sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt
sodium;2-methylbutan-2-olate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0206

TSCA : Yes

Documents