missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tert-pentoxide, 30% in 2-MeTHF, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC252531000

 View more versions of this product

Catalog No. AC252531000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Sodium tert-pentoxide
−11°C
100mL
C5H11NaO
2-butanol, 2-methyl-, sodium salt, sodium 2-methylbutan-2-olate, sodium t-amylate, sodium tert-amoxide, sodium tert-amylate, sodium tert-pentoxide, sodium-t-amylate, sodium-tert-pentoxide, tert-amyl alcohol sodium salt, unii-51z39py5z8
CCC(C)(C)[O-].[Na+]
110.13
110.13
0.9100g/mL
0.91
14593-46-5,96-47-9
NaOC(CH3)2C2H5
CGRKYEALWSRNJS-UHFFFAOYSA-N
sodium;2-methylbutan-2-olate
2733478

Chemical Identifiers

14593-46-5
110.13
2-butanol, 2-methyl-, sodium salt, sodium 2-methylbutan-2-olate, sodium t-amylate, sodium tert-amoxide, sodium tert-amylate, sodium tert-pentoxide, sodium-t-amylate, sodium-tert-pentoxide, tert-amyl alcohol sodium salt, unii-51z39py5z8
sodium;2-methylbutan-2-olate
C5H11NaO
CGRKYEALWSRNJS-UHFFFAOYSA-N
2733478
CCC(C)(C)[O-].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.
May form explosive peroxides.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 238-639-4

SDS
Documents