missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tert-pentoxide, 95%, pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 327555000

 View more versions of this product

Catalog No. AC327555000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
110.13
Cream or light tan
Glass bottle
C5H11NaO
14593-46-5
MFCD00064245
Powder
500g
Pure
Sodium tert-pentoxide
CH3CH2C(CH3)2ONa
95%
95%

Chemical Identifiers

14593-46-5
C5H11NaO
110.13
Sodium tert-pentoxide, 95%
Pure
95%
CH3CH2C(CH3)2ONa
MFCD00064245
≥94%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Self-heating; may catch fire.
Reacts violently with water.

GHS P Statement:
Keep cool. Protect from sunlight.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 238-639-4

TSCA : TSCA