Please login to your online account to display your discounted pricing

Sodium tert-pentoxide, 95%, pure, ACROS Organics™

$86.65 - $86.65

Chemical Identifiers

Chemical Name or Material Sodium tert-pentoxide
Name Note 95%
CAS 14593-46-5
Purity Grade Pure
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C5H11NaO
Linear Formula CH3CH2C(CH3)2ONa
Formula Weight 110.13
MDL Number MFCD00064245
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC327555000

  SDS
thermo fisher scientific
327555000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC327551000

  SDS
thermo fisher scientific
327551000
Glass bottle 100g Each for $86.65
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 110.13
Quantity 500g
Color Cream or light tan
TSCA TSCA
EINECS Number 238-639-4
Purity Grade Pure
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Keep cool. Protect from sunlight.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Self-heating; may catch fire.
Reacts violently with water.
Chemical Name or Material Sodium tert-pentoxide
Molecular Formula C5H11NaO
Linear Formula CH3CH2C(CH3)2ONa
CAS 14593-46-5
Name Note 95%
MDL Number MFCD00064245
Assay Percent Range 95%
Safety and Handling

Danger

GHS P Statement:
Keep cool. Protect from sunlight.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Self-heating; may catch fire.
Reacts violently with water.