missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tert-pentoxide, 30% solution in THF, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC390158000

 View more versions of this product

Catalog No. AC390158000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

110.13
AcroSeal™ Glass Bottle
C5H11NaO
Yellow to light brown solution
Sodium tert-pentoxide
29 to 31%
800mL
0.9200g/mL
MFCD00064245
CH3CH2C(CH3)2ONa
109-99-9,14593-46-5

Chemical Identifiers

14593-46-5,109-99-9
CH3CH2C(CH3)2ONa
MFCD00064245
C5H11NaO
110.13
Sodium tert-pentoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger