Please login to your online account to display your discounted pricing

Sodium tert-pentoxide, 30% solution in THF, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Sodium tert-pentoxide
CAS 14593-46-5
CAS 109-99-9
Molecular Formula C5H11NaO
Linear Formula CH3CH2C(CH3)2ONa
Formula Weight 110.13
MDL Number MFCD00064245
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390151000

  SDS
thermo fisher scientific
AC390151000
AcroSeal™ Glass Bottle in Tin Can 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC390158000

  SDS
thermo fisher scientific
AC390158000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 110.13
Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 0.9200g/mL
Chemical Name or Material Sodium tert-pentoxide
Molecular Formula C5H11NaO
Linear Formula CH3CH2C(CH3)2ONa
Appearance of Solution Yellow to light brown solution
MDL Number MFCD00064245
CAS 14593-46-5
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. Reacts violently with water. May form explosive peroxides.
CAS 109-99-9
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection
Concentration 29 to 31%
Safety and Handling

Danger

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection

GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. Reacts violently with water. May form explosive peroxides.