missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tert-butoxide, 99%, pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 252551000

 View more versions of this product

Catalog No. AC252551000



Description

Description

Specifications

Specifications

Sodium tert-butoxide
Glass bottle
100g
99%
NaOC(CH3)3
16,55
Solubility in water: reacts.
CC(C)(C)[O-].[Na+]
96.1
96.1
Pure
12°C
180.0°C
865-48-5
C4H9NaO
MFCD00040575
2-propanol, 2-methyl-, sodium salt, naotbu, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium tert-butoxide, sodium-t-butoxide, sodium-tert-butoxide, t-butoxy sodium, tbuona, tert-butoxysodium
MFRIHAYPQRLWNB-UHFFFAOYSA-N
sodium;2-methylpropan-2-olate
23676156
99%

Chemical Identifiers

865-48-5
96.1
MFRIHAYPQRLWNB-UHFFFAOYSA-N
23676156
CC(C)(C)[O-].[Na+]
C4H9NaO
MFCD00040575
2-propanol, 2-methyl-, sodium salt, naotbu, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium tert-butoxide, sodium-t-butoxide, sodium-tert-butoxide, t-butoxy sodium, tbuona, tert-butoxysodium
sodium;2-methylpropan-2-olate
Safety and Handling

Safety and Handling

GHS H Statement
Self-heating; may catch fire.
Causes severe skin burns and eye damage.
Reacts violently with water.
Flammable solid.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open f

GHS Signal Word: Danger

EINECSNumber : 212-741-9

SDS Documents