missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium tellurite(IV), 97%

Manufacturer:  Alfa Aesar™ A156330B

 View more versions of this product

Catalog No. AAA156330B



Specifications

Specifications

Sodium tellurite(IV)
Na2O3Te
MFCD00036141
14,8685
VOADVZVYWFSHSM-UHFFFAOYSA-L
disodium;tellurite
24935
97%
10102-20-2
Na2TeO3
UN3284
sodium tellurite, disodium tellurite, sodium tellurate iv, disodium trioxotellurate, unii-0bb57la23y, tellurous acid, disodium salt, sodium tellurite iv, telluric acid h2teo3 , disodium salt, telluric acid, disodium salt, sodium tellurite iv
[O-][Te](=O)[O-].[Na+].[Na+]
221.577
221.58
1000g

Chemical Identifiers

10102-20-2
221.577
VOADVZVYWFSHSM-UHFFFAOYSA-L
24935
[O-][Te](=O)[O-].[Na+].[Na+]
Na2O3Te
MFCD00036141
sodium tellurite, disodium tellurite, sodium tellurate iv, disodium trioxotellurate, unii-0bb57la23y, tellurous acid, disodium salt, sodium tellurite iv, telluric acid h2teo3 , disodium salt, telluric acid, disodium salt, sodium tellurite iv
disodium;tellurite
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TELLURIUM COMPOUND, N.O.S.

RTECSNumber : WY2450000

TSCA : TSCA

Documents