Learn More
Sodium tellurate(VI) hydrate, Te 45% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 07510618
Description
Sodium tellurite is an intermediate in the extraction of the element, tellurium; it is a product obtained from anode slimes and is a precursor to tellurium. Solutions of sodium tellurite are used for black or blue-black coatings on iron, steel, aluminum, and copper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium tellurate (VI) hydrate | |
10101-83-4 | |
50 g | |
MFCD00014242 | |
sodium tellurate, unii-7kar02f0r0, natriumtellurat, 2na.o4te, disodium tellurate, telluric acid h2teo4 , sodium salt 1:2, telluric acid h2teo4 ,disodium salt, dihydrate 8ci,9ci | |
XERQTZLDFHNZIC-UHFFFAOYSA-L | |
disodium;tellurate | |
13652393 | |
Te 45% min |
(decomposition) | |
Crystalline | |
Na2O4Te | |
14,8686 | |
Soluble in water. | |
[Na+].[Na+].[O-][Te]([O-])(=O)=O | |
237.58 | |
237.58 (Anhydrous) |
Chemical Identifiers
10101-83-4 | |
237.58 | |
XERQTZLDFHNZIC-UHFFFAOYSA-L | |
13652393 |
Na2O4Te | |
MFCD00014242 | |
sodium tellurate, unii-7kar02f0r0, natriumtellurat, 2na.o4te, disodium tellurate, telluric acid h2teo4 , sodium salt 1:2, telluric acid h2teo4 ,disodium salt, dihydrate 8ci,9ci | |
[Na+].[Na+].[O-][Te]([O-])(=O)=O |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 233-259-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only