missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium taurocholate hydrate, 97%, Alfa Aesar

Manufacturer:  Alfa Aesar A1834606

 View more versions of this product

Catalog No. AAA1834606


 

Description

Description

Applications
Sodium taurocholate is a bile acid having detergent capabilities; natural surfactant. It is used for solubilization of protein. In the preparation of digestive enzymes, it is used as an emulsifier.

Solubility
Soluble in Water (50 mg/mL - Clear Solution)

Notes
Stable under normal storage conditions. It is incompatible with moisture and strong oxidizing agents.
Specifications

Specifications

Sodium Taurocholate Hydrate
0.97
JAJWGJBVLPIOOH-VXFFEJGCSA-M
sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
131632374
5g
145-42-6
C26H44NNaO7S
CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]
537.688
537.73 (Anhydrous)

Chemical Identifiers

145-42-6
537.688
131632374
C26H44NNaO7S
JAJWGJBVLPIOOH-VXFFEJGCSA-M
SDS Documents