missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Tartrate, For Nickel, 10% (w/w), LabChem™

$118.23 - $118.23

Chemical Identifiers

CAS 7732-18-5,6106-24-7
CAS Max % 90,10
Linear Formula Na2C4H4O6·2H2O
Formula Weight 230.08
Chemical Name or Material Sodium Tartrate, For Nickel
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC249501 Lab Chem
LC249501
500mL Each for $118.23
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,6106-24-7
Na2C4H4O6·2H2O
Sodium Tartrate, For Nickel
90,10
230.08

Specifications

7732-18-5,6106-24-7
Liquid
230.08
Passes Test
Soluble in water
Room Temperature
Carbon monoxide; Carbon dioxide
500mL
10% (w/w)
90,10
Na2C4H4O6·2H2O
1.06g/mL
Poly Bottle
1.06g/mL
1.2cSt
Clear, colorless
Sodium Tartrate, For Nickel

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.