Please login to your online account to display your discounted pricing

Sodium Tartrate, Dihydrate, ACS Grade, 99.0 to 101.0%, LabChem™

$185.26 - $185.26

Chemical Identifiers

Chemical Name or Material Sodium Tartrate
CAS 6106-24-7
CAS Max % 100
Name Note Dihydrate
Purity Grade ACS Grade
Linear Formula Na2C4H4O6·2H2O
Formula Weight 230.08
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC249451

 
lab chem
LC249451
500g Each for $185.26
Description & Specifications

Specifications

Formula Weight 230.08
CAS 6106-24-7
Packaging Poly Bottle
Linear Formula Na2C4H4O6·2H2O
Quantity 500g
Purity Grade ACS Grade
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Color White
Melting Point 150°C
Solubility Soluble in water
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Storage Information Room Temperature
Identification Passes Test
Chemical Name or Material Sodium Tartrate
Physical Form Crystals
Name Note Dihydrate
CAS Max % 100
Health Hazard 2 GHS H Statement
Product is not hazardous.
Assay 99.0 to 101.0%
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.