missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Tartrate ACS AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 6106-24-7
Physical Form Granular
Molecular Formula NaOCO(CHOH)2COONa·2H2O
Formula Weight 230.08
Chemical Name or Material Sodium Tartrate, Dihydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK2386500 Avantor Performance
238604
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6106-24-7
NaOCO(CHOH)2COONa·2H2O
Sodium Tartrate, Dihydrate
Granular
230.08
ACS Grade

Specifications

230.08
Granular
6106-24-7
Sodium Tartrate, Dihydrate
500g
NaOCO(CHOH)2COONa·2H2O
POLYSTORMOR
ACS Grade