Please login to your online account to display your discounted pricing

Sodium Tartrate, 10% (w/v) Aqueous Solution, Ricca Chemical


Quantity 100mL
Packaging Plastic Bottle

Manufacturer: ricca chemical company  RIR7850000100A

 View more versions of this product

Catalog No. R7850000100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Sodium Tartrate
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 6106-24-7
CAS Min % 8.91
CAS Max % 9.28
Physical Form Liquid
Packaging Plastic Bottle
Color Clear colorless
Quantity 100mL

Specifications

Chemical Name or Material Sodium Tartrate
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 6106-24-7
CAS Min % 8.91
CAS Max % 9.28
Physical Form Liquid