missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Sulfite, For Lead, 5% (w/v), LabChem™

$82.91 - $82.91

Chemical Identifiers

CAS 7732-18-5,7757-83-7
CAS Max % 95,5
Linear Formula Na2SO3
Formula Weight 126.04
Chemical Name or Material Sodium Sulfite, For Lead
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC249401 Lab Chem
LC249401
500mL Each for $82.91
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7757-83-7
Na2SO3
Sodium Sulfite, For Lead
95,5
126.04

Specifications

126.04
Soluble in water
500mL
7757-83-7,7732-18-5
Liquid
Room Temperature
Sodium Sulfite, For Lead
Na2SO3
Clear, colorless
Poly Bottle
Sulfur compounds
95,5
Passes Test
5% (w/v)

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.