missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Sulfite, Anhydrous 97%, ACROS Organics™

Manufacturer:  Acros Organics AC196630100

 View more versions of this product

Catalog No. AC196630100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Reduction reagent
Specifications

Specifications

Sodium sulfite
97%
Na2SO3
15,8811
Solubility in water: 230g/mL (20°C). Other solubilities: soluble in glycerol,ppractically insoluble in alcohol
[O-]S(=O)[O-].[Na+].[Na+]
126.04
CHEBI:86477
Fine Crystals or Crystalline Powder
Extra Pure
10ppm max.
Plastic drum
Solubility in water: 230 g/mL (20°C). Other solubilities: soluble in glycerol, ppractically insoluble in alcohol
White to Yellow
8.5 to 10.5 (5% soln.)
7757-83-7
Na2O3S
02,387
disodium sulfite
GEHJYWRUCIMESM-UHFFFAOYSA-L
disodium;sulfite
24437
126.04
97%
2.6300g/mL
10ppm max.
50ppm max.
2.63
>500.0°C
10kg

Chemical Identifiers

7757-83-7
126.04
disodium sulfite
CHEBI:86477
[O-]S(=O)[O-].[Na+].[Na+]
Na2O3S
GEHJYWRUCIMESM-UHFFFAOYSA-L
24437
disodium;sulfite
SDS Documents