Please login to your online account to display your discounted pricing

Sodium Sulfite, ACS Grade, ≥ 98.0%, LabChem™

$49.51 - $49.51

Chemical Identifiers

Chemical Name or Material Sodium Sulfite
CAS 7757-83-7
CAS Max % 100
Purity Grade ACS Grade
Linear Formula Na2SO3
Formula Weight 126.04
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC249301

 
lab chem
LC249301
500g Each for $49.51
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥98.0%
Formula Weight 126.04
Chemical Name or Material Sodium Sulfite
EINECS Number 231-821-4
Identification Passes Test
Alkalinity ≤0.03meq/gcSt
Packaging Poly Bottle
Purity Grade ACS Grade
Solubility Soluble in water
Specific Gravity 2.63g/cm3
Storage Information Room Temperature
Linear Formula Na2SO3
Acidity Passes Test
CAS 7757-83-7
Decomposition Information Sodium oxide; Sulfur compounds
Color White
Melting Point 500°C
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Density 2.63g/cm3
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.