missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Sodium Sulfite ACS AR Granular, Macron Fine Chemicals™
macron
Chemical Identifiers
CAS |
7757-83-7 |
Molecular Formula |
Na2O3S |
Molecular Weight (g/mol) |
126.037 |
InChI Key |
GEHJYWRUCIMESM-UHFFFAOYSA-L |
Synonym |
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit |
PubChem CID |
24437 |
ChEBI |
CHEBI:86477 |
IUPAC Name |
disodium;sulfite |
SMILES |
[O-]S(=O)[O-].[Na+].[Na+] |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
7757-83-7 |
|
126.037 |
|
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit |
|
CHEBI:86477 |
|
[O-]S(=O)[O-].[Na+].[Na+] |
|
Na2O3S |
|
GEHJYWRUCIMESM-UHFFFAOYSA-L |
|
24437 |
|
disodium;sulfite |
Specifications
|
7757-83-7 |
|
Na2O3S |
|
GEHJYWRUCIMESM-UHFFFAOYSA-L |
|
disodium;sulfite |
|
24437 |
|
126.04 |
|
POLYSTORMOR |
|
500g |
|
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit |
|
[O-]S(=O)[O-].[Na+].[Na+] |
|
126.037 |
|
CHEBI:86477 |
|
ACS |
|
Sodium Sulfite, Anhydrous |
SDS
missing translation for 'documents'