Learn More
Sodium sulfite, 98.5%, for analysis, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 219272500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Sodium sulfite | |
Anhydrous, For analysis | |
White | |
250 g | |
Na2O3S | |
MFCD00003503 | |
15, 8811 | |
Solubility in water: 230g/mL (20°C). Other solubilities: soluble in glycerol,ppractically insoluble in alcohol | |
[O-]S(=O)[O-].[Na+].[Na+] | |
126.04 | |
CHEBI:86477 | |
98.5% | |
Plastic bottle | |
2.6300g/mL |
>500.0°C | |
7757-83-7 | |
Fine Crystals or Crystalline Powder | |
98% min. (Iodimetry) | |
Na2SO3 | |
02,387 | |
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit | |
GEHJYWRUCIMESM-UHFFFAOYSA-L | |
disodium;sulfite | |
24437 | |
126.04 | |
Analytical | |
2.63 |
Chemical Identifiers
7757-83-7 | |
126.04 | |
GEHJYWRUCIMESM-UHFFFAOYSA-L | |
24437 | |
disodium;sulfite |
Na2O3S | |
MFCD00003503 | |
sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit | |
CHEBI:86477 | |
[O-]S(=O)[O-].[Na+].[Na+] |
Safety and Handling
GHS H Statement:
Contact with acids liberates toxic gas.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 231-821-4
RUO – Research Use Only