Please login to your online account to display your discounted pricing

Sodium Sulfite, 5% (w/v) Aqueous Solution, Ricca Chemical

$49.21 - $118.09

Chemical Identifiers

Chemical Name or Material Sodium Sulfite
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7757-83-7
CAS Min % 4.67
CAS Max % 4.86
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7580-16

 
ricca chemical company
RI758016
Poly natural 500mL Each for $49.21

7580-32

 
ricca chemical company
RI758032
Poly natural 1L Each for $118.09
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Concentration or Composition Notes 5% (w/v)
Solvent or Matrix Aqueous Solution
CAS Max % 97.0
CAS Min % 93.0
CAS 7732-18-5
Chemical Name or Material Sodium Sulfite
CAS 7757-83-7
CAS Min % 4.67
CAS Max % 4.86
Packaging Poly natural