missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Sulfite, 2.5% (w/v), LabChem™

$68.91 - $68.91

Chemical Identifiers

CAS 7732-18-5,7757-83-7
CAS Max % 97.5,2.5
Linear Formula Na2SO3
Formula Weight 126.04
Chemical Name or Material Sodium Sulfite
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC249352 Lab Chem
LC249352
1L Each for $68.91
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7757-83-7
Na2SO3
Sodium Sulfite
97.5,2.5
126.04

Specifications

126.04
97.5,2.5
Liquid
Na2SO3
Clear, colorless
Soluble in water
Sodium Sulfite
Sulfur compounds
7732-18-5,7757-83-7
Room Temperature
Passes Test
1L
Poly Bottle
2.5% (w/v)

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.