Please login to your online account to display your discounted pricing

Sodium Sulfite, 1.00 Molar, 2.00 Normal, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Sulfite
Name Note 1.00 Molar, 2.00 Normal
CAS 7757-83-7
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 1.000 ± 0.001 M at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R783200020F

 
ricca chemical company
RIR783200020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R7832000500

 
ricca chemical company
RIR7832000500C
Amber glass 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sodium Sulfite
Name Note 1.00 Molar, 2.00 Normal
CAS 7757-83-7
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 1.000 ± 0.001 M at 20°C
Odor Odorless
Packaging Cubitainer
Solubility Solubility in Water: Infinite
Solvent or Matrix Aqueous Solution
Color Clear colorless
Quantity 20L