Please login to your online account to display your discounted pricing

Sodium sulfide nonahydrate, extra pure, ACROS Organics™

$102.21 - $102.21

Chemical Identifiers

Chemical Name or Material Sodium sulfide nonahydrate
CAS 1313-84-4
Purity Grade Extra Pure
Assay 32.0 to 38.0% (Na2S)
Molecular Formula Na29H2O
Linear Formula Na2S·9H2O
Formula Weight 240.18
MDL Number MFCD00149184
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC387065000

  SDS
thermo fisher scientific
AC387065000
Glass bottle 500g Each for $102.21
Description & Specifications

Specifications

Purity Grade Extra Pure
Physical Form Crystalline Chunks
Formula Weight 240.18
Quantity 500g
Color Colorless or slightly yellow
Merck Index 15, 8810
Assay 32.0 to 38.0% (Na2S)
Linear Formula Na2S·9H2O
Chemical Name or Material Sodium sulfide nonahydrate
Packaging Glass bottle
Molecular Formula Na29H2O
CAS 1313-84-4
Thiosulphate 5000ppm max.
Total Nitrogen 0.005% max.
Trace Analysis 2000ppm max.
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May be corrosive to metals.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Keep only in original container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00149184
Safety and Handling

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Keep only in original container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May be corrosive to metals.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.