Please login to your online account to display your discounted pricing

Sodium Sulfide Nonahydrate (Crystalline/Certified ACS), Fisher Chemical


This chemical is deliquescent

Quantity 2.5kg
Packaging Glass Bottle

Manufacturer: fisher scientific company  S425212

 View more versions of this product

Catalog No. S425-212

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Purity Grade Certified ACS Grade
Formula Weight 240.18
Melting Point 50°C
Quantity 2.5kg
DOT Information DOT Class 8, : Corrosive
Physical Form Solid
Molecular Formula Na2S · 9 H2O
Viscosity [NULL]
Color Light Yellow
Chemical Name or Material Sodium Sulfide Nonahydrate
Identification Pass Test
Iron (Fe) Pass Test
Packaging Glass Bottle
Assay Percent Range ≥98.0 %
MDL Number MFCD00149184
CAS 1313-84-4
Ammonium 0.01% max.
Sulfite or Thiosulfate 0.1% max.
ChemAlert Storage Symbol White
Synonym Sodium Monosulfide
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Toxic in contact with skin. Causes burns by all exposure routes. Contact with acids liberates toxic gas. Very toxic to aquatic organisms. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:1
Instability:1

Specifications

Chemical Name or Material Sodium Sulfide Nonahydrate
CAS 1313-84-4
Purity Grade Certified ACS Grade
Physical Form Solid
Assay ≥98.0%
Assay Percent Range ≥98.0 %
Molecular Formula Na2S · 9 H2O
Formula Weight 240.18
MDL Number MFCD00149184
Synonym Sodium Monosulfide
Accessories
 

Emergency Overview
Toxic in contact with skin. Causes burns by all exposure routes. Contact with acids liberates toxic gas. Very toxic to aquatic organisms. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:1
Instability:1

DANGER!