missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Sulfide Nonahydrate (Crystalline/Certified ACS), Fisher Chemical

This chemical is deliquescent

$335.92 - $1,648.38

Chemical Identifiers

CAS 1313-84-4
Physical Form Solid
Assay Percent Range ≥98.0 %
Molecular Formula Na2S · 9 H2O
Formula Weight 240.18
MDL Number MFCD00149184
Synonym Sodium Monosulfide
Chemical Name or Material Sodium Sulfide Nonahydrate
Assay ≥98.0%
Grade Certified ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
S425-212 SDS Fisher Scientific Company
S425212
Glass Bottle 2.5kg Each for $1,648.38
Case of 4 Each for $6,593.52
S425-500 SDS Fisher Scientific Company
S425500
Poly Bottle 500g Each for $335.92
Case of 6 Each for $2,015.52
Specifications Description & Specifications

Chemical Identifiers

1313-84-4
≥98.0 %
240.18
Sodium Monosulfide
≥98.0%
Solid
Na2S · 9 H2O
MFCD00149184
Sodium Sulfide Nonahydrate
Certified ACS Grade

Specifications

1313-84-4
≥98.0 %
0.01% max.
MFCD00149184
Pass Test
Glass Bottle
Light Yellow
2.5kg
≥98.0%
Solid
Na2S · 9 H2O
240.18
Sodium Monosulfide
Pass Test
0.1% max.
50°C
Sodium Sulfide Nonahydrate
Certified ACS Grade

Safety and Handling

DANGER!

Emergency Overview
Toxic in contact with skin. Causes burns by all exposure routes. Contact with acids liberates toxic gas. Very toxic to aquatic organisms. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:1
Instability:1

DOTInformation : DOT Class 8, : Corrosive